Assay Details
Target Gene Details
Entrez Gene ID: | 242316 |
Gene Name: | growth differentiation factor 6 |
Gene Aliases: |
BMP-13, BMP13, GDF16 |
Location: |
Chr.4:9844372-9862345 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gdf6 | NM_013526.1 | 2 | 3026 | NP_038554.1 |
| AJ537425.1 | 2 | 3026 | CAD60935.1 |
More Information
Set Membership: |
|

Genomic Map