Assay Details
Target Gene Details
Entrez Gene ID: | 14579 |
Gene Name: | GTP binding protein (gene overexpressed in skeletal muscle) |
Gene Aliases: |
AV020497 |
Location: |
Chr.4:11704447-11714993 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gem | NM_010276.4 | 2 | 339 | NP_034406.2 |
| AK153999.1 | 1 | 1669 | BAE32306.1 | |
| AK157655.1 | 2 | 341 | ||
| AK158393.1 | 2 | 341 | BAE34484.1 | |
| AK168401.1 | 2 | 340 | BAE40322.1 | |
| AK169699.1 | 1 | 1669 | ||
| AK171336.1 | 2 | 277 | BAE42399.1 | |
| BC029668.1 | 2 | 315 | AAH29668.1 | |
| BY133754.1 | ||||
| U10551.1 | 1 | 317 | AAA64912.1 | |
| U13053.1 | 2 | 330 | AAC52145.1 |

Genomic Map