Assay Details
Target Gene Details
Entrez Gene ID: | 231050 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 11 |
Gene Aliases: |
A430075I06Rik, AI648252, E430002F06Rik |
Location: |
Chr.5:25222893-25265918 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galnt11 | NM_144908.3 | 12 | 1986 | NP_659157.1 |
| AF495528.1 | 11 | 1858 | AAQ06668.1 | |
| AK088014.1 | 12 | 1975 | BAC40096.1 | |
| AK161524.1 | 12 | 1896 | BAE36442.1 | |
| BC011428.1 | 12 | 1962 | AAH11428.1 | |
| BC021504.1 | 12 | 1987 | AAH21504.1 | |
| BC034184.1 | 11 | 1886 | AAH34184.1 | |
| BC034185.1 | 11 | 1938 | AAH34185.1 | |
| BC036143.1 | 11 | 1887 | AAH36143.1 | |
| BC036145.1 | 11 | 1880 | AAH36145.1 | |
| Y12435.1 | CAC79626.1 |
More Information
Set Membership: |
|

Genomic Map