Assay Details
Target Gene Details
Entrez Gene ID: | 625464 |
Gene Name: | predicted gene 6588 |
Gene Aliases: |
EG625464 |
Location: |
Chr.5:112449426-112451738 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm6588 | NM_001177504.1 | 1 | 771 | NP_001170975.1 |
| AK133454.1 | 1 | 771 | BAE21665.1 |
Target Gene Details
Entrez Gene ID: | 56747 |
Gene Name: | seizure related 6 homolog like |
Gene Aliases: |
AI843918, AIG1, AW539724, Acig1, BSRP-B, mKIAA0927 |
Location: |
Chr.5:112419151-112578042 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sez6l | NM_001253916.1 | NP_001240845.1 | ||
| NM_001253917.1 | NP_001240846.1 | |||
| NM_019982.3 | NP_064366.2 | |||
| AB206790.1 | BAE44443.1 | |||
| AK083229.1 | BAC38818.1 | |||
| AK122401.1 | BAC65683.1 | |||
| BC056471.1 | AAH56471.1 | |||
| BC065117.1 | AAH65117.1 | |||
| BC065129.1 | AAH65129.1 | |||
| BC094376.1 | AAH94376.1 | |||
| BC099839.1 | AAH99839.1 | |||
| DQ167195.1 |
More Information
Set Membership: |
|

Genomic Map