Assay Details
Target Gene Details
Entrez Gene ID: | 56222 |
Gene Name: | Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4 |
Gene Aliases: |
MRG-2, Mrg2 |
Location: |
Chr.4:120666563-120667820 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cited4 | NM_019563.2 | 1 | 835 | NP_062509.1 |
| AF143369.1 | 1 | 805 | AAD37357.1 | |
| AK012136.1 | 1 | 827 | BAB28054.1 | |
| AK154030.1 | 1 | 826 | BAE32327.1 | |
| BC025116.1 | 2 | 507 | AAH25116.1 | |
| BC080723.1 | 2 | 592 | AAH80723.1 | |
| BC132360.1 | 1 | 782 | ||
| BC132362.1 | 1 | 782 |
More Information
Set Membership: |
|

Genomic Map