Assay Details
Target Gene Details
Entrez Gene ID: | 15569 |
Gene Name: | ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B) |
Gene Aliases: |
Hub, mel-N1 |
Location: |
Chr.4:91250766-91400064 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elavl2 | NM_001177883.1 | 7 | 3665 | NP_001171354.1 |
| NM_010486.2 | 7 | 3263 | NP_034616.1 | |
| NM_207685.1 | 8 | 3372 | NP_997568.1 | |
| NM_207686.1 | 6 | 3224 | NP_997569.1 | |
| AK014085.1 | 1 | 1294 | ||
| BC046598.1 | 6 | 3148 | AAH46598.2 | |
| BC049125.1 | 8 | 3372 | AAH49125.1 | |
| BC058393.1 | 8 | 3372 | AAH58393.1 |
More Information
Set Membership: |
|

Genomic Map