Assay Details
Target Gene Details
Entrez Gene ID: | 66717 |
Gene Name: | coiled-coil domain containing 96 |
Gene Aliases: |
4921513E08Rik |
Location: |
Chr.5:36484588-36488171 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc96 | NM_025725.2 | 1 | 2982 | NP_080001.1 |
| AK030023.1 | 1 | 2982 | BAC26742.1 | |
| BC125034.1 |

Genomic Map