Assay Details
Target Gene Details
Entrez Gene ID: | 12425 |
Gene Name: | cholecystokinin A receptor |
Gene Aliases: |
AW106902 |
Location: |
Chr.5:53698485-53707704 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cckar | NM_009827.2 | 3 | 888 | NP_033957.1 |
| AK004730.2 | 3 | 746 | BAB23512.1 | |
| AK143895.1 | 3 | 736 | ||
| AK146738.1 | 3 | 889 | ||
| AK147052.1 | 3 | 889 | ||
| BC020534.1 | 3 | 708 | AAH20534.1 |
More Information
Set Membership: |
|

Genomic Map