Assay Details
Target Gene Details
Entrez Gene ID: | 384281 |
Gene Name: | glutamyl-tRNA(Gln) amidotransferase, subunit C |
Gene Aliases: |
2010003O18Rik |
Location: |
Chr.5:115333242-115341161 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gatc | NM_029645.3 | 3 | 653 | NP_083921.1 |
| AK008080.1 | 3 | 658 | BAB25446.1 | |
| AK034326.1 | 3 | 653 | BAC28675.1 | |
| BC021775.1 | 3 | 646 | AAH21775.1 |

Genomic Map