Assay Details
Target Gene Details
Entrez Gene ID: | 14467 |
Gene Name: | glioblastoma amplified sequence |
Gene Aliases: |
AV006093, Nipsnap2 |
Location: |
Chr.5:129725044-129758327 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gbas | NM_008095.4 | 10 | 1062 | NP_032121.3 |
| AJ001261.1 | CAA04635.1 | |||
| AK170336.1 | 10 | 1008 | BAE41726.1 | |
| AK183347.1 | ||||
| AK187878.1 | ||||
| BC055893.1 | 10 | 1011 | AAH55893.1 |

Genomic Map