Assay Details
Target Gene Details
Entrez Gene ID: | 54325 |
Gene Name: | elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 1 |
Gene Aliases: |
AA407424, BB151133, Ssc1 |
Location: |
Chr.4:118428081-118432953 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elovl1 | NM_001039175.2 | 7 | 1412 | NP_001034264.1 |
| NM_001039176.2 | 8 | 1610 | NP_001034265.1 | |
| NM_019422.3 | 8 | 1603 | NP_062295.1 | |
| BM445297.1 | 1 | 357 | ||
| BY689799.1 | 1 | 175 |
More Information
Set Membership: |
|

Genomic Map