Assay Details
Target Gene Details
Entrez Gene ID: | 68943 |
Gene Name: | PTEN induced putative kinase 1 |
Gene Aliases: |
1190006F07Rik, AU042772, AW557854, BRPK, mFLJ00387 |
Location: |
Chr.4:138313409-138326296 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pink1 | NM_026880.2 | 8 | 1828 | NP_081156.2 |
| AB053476.1 | 8 | 1760 | BAB55651.1 | |
| AF316872.1 | 8 | 1806 | AAK28061.1 | |
| AK004507.1 | 4 | 676 | ||
| AK050322.1 | 8 | 1808 | ||
| AK155474.1 | 8 | 1814 | BAE33284.1 | |
| AK165480.1 | 8 | 1806 | ||
| AK179666.1 | 1 | 53 | ||
| AK181855.1 | 2 | 365 | ||
| AK192353.1 | 1 | 240 | ||
| AK195623.1 | ||||
| AK204507.1 | 1 | 240 | ||
| AK212959.1 | 1 | 240 | ||
| AK213673.1 | 1 | 240 | ||
| AK219274.1 | 1 | 44 | ||
| AK220318.1 | 8 | 1782 | BAD90392.1 | |
| BC044743.1 | 3 | 524 | AAH44743.1 | |
| BC054349.1 | 4 | 806 | AAH54349.1 | |
| BC067066.1 | 8 | 1796 | AAH67066.1 | |
| BC069968.1 | AAH69968.1 |
More Information
Set Membership: |
|

Genomic Map