Assay Details
Target Gene Details
Entrez Gene ID: | 11769 |
Gene Name: | adaptor protein complex AP-1, sigma 1 |
Gene Aliases: |
AP19 |
Location: |
Chr.5:137034993-137046060 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap1s1 | NM_007457.2 | 5 | 746 | NP_031483.1 |
| AK002225.1 | 5 | 724 | BAB21947.1 | |
| AK196123.1 | ||||
| BC052692.1 | 5 | 750 | AAH52692.1 | |
| M62418.1 | 5 | 746 | AAA37243.1 |
More Information
Set Membership: |
|

Genomic Map