Assay Details
Target Gene Details
Entrez Gene ID: | 12757 |
Gene Name: | clathrin, light polypeptide (Lca) |
Gene Aliases: |
AV026556, Lca |
Location: |
Chr.4:44003816-44032848 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clta | NM_001080384.1 | 5 | 992 | NP_001073853.1 |
| NM_001080385.1 | 7 | 1082 | NP_001073854.1 | |
| NM_001080386.1 | 5 | 986 | NP_001073855.1 | |
| NM_001290470.1 | 6 | 1021 | NP_001277399.1 | |
| NM_016760.2 | 6 | 1046 | NP_058040.2 | |
| AI467006.1 | 1 | 118 | ||
| AK146965.1 | 6 | 1625 | BAE27571.1 | |
| AK149761.1 | 5 | 958 | BAE29067.1 | |
| AK150148.1 | 5 | 958 | BAE29342.1 | |
| AK152397.1 | 5 | 962 | BAE31185.1 | |
| AK168130.1 | 5 | 986 | BAE40099.1 | |
| AK205683.1 | 1 | 124 | ||
| AK207054.1 | 1 | 124 | ||
| AK207565.1 | 1 | 124 | ||
| AK209788.1 | 1 | 124 | ||
| AK209982.1 | 1 | 124 | ||
| AK214893.1 | 1 | 124 | ||
| BC057660.1 | 5 | 952 | AAH57660.1 | |
| BE333066.1 | 1 | 108 | ||
| U91848.1 | 6 | 1002 | AAB51286.1 |
More Information
Set Membership: |
|

Genomic Map