Assay Details
Target Gene Details
Entrez Gene ID: | 68053 |
Gene Name: | UBX domain protein 2B |
Gene Aliases: |
3110003A22Rik, 6430407D20Rik, AI451665, p37 |
Location: |
Chr.4:6191105-6219788 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubxn2b | NM_026534.2 | 8 | 2154 | NP_080810.2 |
| AK032332.1 | 8 | 2154 | BAC27819.1 | |
| AK049018.1 | 8 | 2162 | BAC33515.1 | |
| BC076632.1 | 8 | 2154 | AAH76632.1 |
More Information
Set Membership: |
|

Genomic Map