Assay Details
Target Gene Details
Entrez Gene ID: | 242521 |
Gene Name: | kelch-like 9 |
Gene Aliases: |
8030469P05, C530050O22Rik, ENSMUSG00000070923, mKIAA1354 |
Location: |
Chr.4:88718292-88722508 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhl9 | NM_172871.2 | 1 | 3517 | NP_766459.2 |
| AK129338.1 | 1 | 3419 | BAC98148.1 |

Genomic Map