Assay Details
Target Gene Details
Entrez Gene ID: | 26562 |
Gene Name: | neurochondrin |
Gene Aliases: |
AU042419, MMS10-AE, Ms10ae, mKIAA0607, norbin |
Location: |
Chr.4:126743750-126753566 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncdn | NM_011986.4 | 7 | 3453 | NP_036116.3 |
| AK122327.1 | 5 | 2798 | BAC65609.1 | |
| AK154889.1 | 7 | 3434 | BAE32905.1 | |
| AK170502.1 | 8 | 3168 | BAE41841.1 | |
| AK185185.1 | ||||
| BC017126.1 | 7 | 3384 | AAH17126.1 | |
| BE985399.1 | 1 | 266 |
More Information
Set Membership: |
|

Genomic Map