Assay Details
Target Gene Details
Entrez Gene ID: | 67916 |
Gene Name: | phosphatidic acid phosphatase type 2B |
Gene Aliases: |
1110003O22Rik, 2610002D05Rik, AV025606, D4Bwg0538e, D4Bwg1535e, Lpp3, Ppab2b |
Location: |
Chr.4:105157347-105232767 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppap2b | NM_080555.2 | 6 | 1605 | NP_542122.1 |
| AK076183.1 | 6 | 1301 | ||
| AK159136.1 | 6 | 1691 | BAE34848.1 | |
| AK160052.1 | ||||
| AK160056.1 | 6 | 1505 | BAE35594.1 | |
| AK163427.1 | 1 | 105 |
More Information
Set Membership: |
|

Genomic Map