Assay Details
Target Gene Details
Entrez Gene ID: | 381605 |
Gene Name: | TBC1 domain family, member 2 |
Gene Aliases: |
A630005A06Rik, BC022630, Gm1037, PARIS-1, PARIS1 |
Location: |
Chr.4:46604390-46650877 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d2 | NM_198664.3 | 13 | 3782 | NP_941066.3 |
| AK148289.1 | 13 | 3698 | ||
| AK211313.1 |
More Information
Set Membership: |
|

Genomic Map