Assay Details
Target Gene Details
Entrez Gene ID: | 381605 |
Gene Name: | TBC1 domain family, member 2 |
Gene Aliases: |
A630005A06Rik, BC022630, Gm1037, PARIS-1, PARIS1 |
Location: |
Chr.4:46604390-46650877 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d2 | NM_198664.3 | 6 | 1403 | NP_941066.3 |
| AK148254.1 | 6 | 1406 | BAE28440.1 | |
| AK148289.1 | 6 | 1320 |
More Information
Set Membership: |
|

Genomic Map