Assay Details
Target Gene Details
Entrez Gene ID: | 269523 |
Gene Name: | valosin containing protein |
Gene Aliases: |
3110001E05, CDC48, p97, p97/VCP |
Location: |
Chr.4:42979964-43000507 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vcp | NM_009503.4 | 1 | 62 | NP_033529.3 |
| AK028264.1 | BAC25849.1 | |||
| AK153012.1 | ||||
| AK153249.1 | BAE31840.1 | |||
| AK159177.1 | BAE34876.1 | |||
| AK159475.1 | ||||
| AK159509.1 | BAE35141.1 | |||
| AK167794.1 | BAE39824.1 | |||
| BC049114.1 | AAH49114.1 | |||
| BY732145.1 | 1 | 62 |

Genomic Map