Assay Details
Target Gene Details
Entrez Gene ID: | 117590 |
Gene Name: | ankyrin repeat and SOCS box-containing 10 |
Gene Aliases: |
Asb-10, BB137407 |
Location: |
Chr.5:24532685-24541047 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb10 | NM_080444.4 | 6 | 2050 | NP_536692.1 |
| AK028538.1 | 6 | 2050 | BAC25997.1 |
More Information
Set Membership: |
|

Genomic Map