Assay Details
Target Gene Details
Entrez Gene ID: | 213948 |
Gene Name: | autophagy related 9B |
Gene Aliases: |
Apg912, Apg9l2, Apgdc2, Gm574, Nos3as, eONE, sONE |
Location: |
Chr.5:24384181-24392143 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atg9b | NM_001002897.3 | 14 | 3606 | NP_001002897.2 |
| AK010032.1 | 2 | 746 | ||
| AY495541.1 | 14 | 3606 | AAS72555.1 | |
| AY515312.1 | 9 | 1779 | AAS87213.1 | |
| AY515313.1 | 14 | 3606 | AAS87214.1 |
Target Gene Details
Entrez Gene ID: | 18127 |
Gene Name: | nitric oxide synthase 3, endothelial cell |
Gene Aliases: |
2310065A03Rik, Nos-3, eNOS, ecNOS |
Location: |
Chr.5:24364819-24384474 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nos3 | NM_008713.4 | NP_032739.3 | ||
| AK077896.1 | BAC37052.1 | |||
| AW489695.1 |
More Information
Set Membership: |
|

Genomic Map