Assay Details
Target Gene Details
Entrez Gene ID: | 243312 |
Gene Name: | leucine rich repeat and fibronectin type III, extracellular 1 |
Gene Aliases: |
A930017N06Rik, AI197272, Ppp1r28 |
Location: |
Chr.5:139907640-139974725 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elfn1 | NM_175522.3 | 2 | 1338 | NP_780731.1 |
| AK044511.1 | 2 | 1338 | BAC31957.1 | |
| BC059029.1 | 1 | 1070 | AAH59029.1 |
More Information
Set Membership: |
|

Genomic Map