Assay Details
Target Gene Details
Entrez Gene ID: | 74155 |
Gene Name: | ERBB receptor feedback inhibitor 1 |
Gene Aliases: |
1300002F13Rik, AI788755, Mig-6, Mig6, RALT |
Location: |
Chr.4:150853801-150868889 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Errfi1 | NM_133753.1 | 4 | 2164 | NP_598514.1 |
| AK004851.1 | 4 | 2183 | ||
| AK139780.1 | 4 | 2180 | ||
| AK166791.1 | 4 | 2183 | BAE39021.1 | |
| AK167156.1 | 4 | 2123 | BAE39297.1 | |
| AK167359.1 | 4 | 2183 | BAE39457.1 | |
| BC005546.1 | 4 | 2164 | AAH05546.1 | |
| BC057646.1 | 4 | 2182 | AAH57646.1 |

Genomic Map