Assay Details
Target Gene Details
Entrez Gene ID: | 242443 |
Gene Name: | glutamate receptor ionotropic, NMDA3A |
Gene Aliases: |
6430537F04, A830097C19Rik, NMDAR-L, NR3A, mKIAA1973 |
Location: |
Chr.4:49661611-49846003 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grin3a | NM_001033351.2 | NP_001028523.1 | ||
| NM_001276355.1 | NP_001263284.1 | |||
| AK138366.1 | ||||
| AK173314.1 | BAD32592.1 |
Target Gene Details
Entrez Gene ID: | 19059 |
Gene Name: | protein phosphatase 3, regulatory subunit B, alpha isoform (calcineurin B, type II) |
Gene Aliases: |
CaNB2, CnB2 |
Location: |
Chr.4:49678747-49681983 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp3r2 | NM_001004025.4 | 1 | 283 | NP_001004025.1 |
| AK029998.1 | 1 | 285 | BAC26725.1 | |
| AK132760.1 | 1 | 308 | BAE21341.1 | |
| AK161263.1 | 1 | 285 | BAE36277.1 | |
| BC100409.1 | 1 | 317 | AAI00410.1 | |
| S43865.1 | 1 | 314 | AAB23172.1 |
More Information
Set Membership: |
|

Genomic Map