Assay Details
Target Gene Details
Entrez Gene ID: | 100213 |
Gene Name: | RUN and SH3 domain containing 2 |
Gene Aliases: |
AI840675, mKIAA0375 |
Location: |
Chr.4:43381982-43427092 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rusc2 | NM_001037709.2 | 2 | 569 | NP_001032798.1 |
| NM_199057.3 | 2 | 530 | NP_951012.2 | |
| AK053542.1 | BAC35421.1 | |||
| AK122263.1 | 2 | 452 | BAC65545.1 | |
| AK132170.1 | 2 | 459 | BAE21009.1 | |
| AK166260.1 | 2 | 472 | ||
| BC050780.1 | 2 | 481 | AAH50780.1 | |
| BC141186.1 | 1 | 303 | ||
| BY237595.1 |

Genomic Map