Assay Details
Target Gene Details
Entrez Gene ID: | 100121 |
Gene Name: | tudor domain containing 7 |
Gene Aliases: |
5730495N10Rik, AI447470, PCTAIRE2BP |
Location: |
Chr.4:45965027-46034766 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tdrd7 | NM_001290475.1 | 15 | 2857 | NP_001277404.1 |
| NM_146142.2 | 15 | 2855 | NP_666254.1 | |
| AK077641.1 | 15 | 2864 | ||
| AK150077.1 | 15 | 2755 | ||
| BC025099.1 | 3 | 511 | AAH25099.1 | |
| BC029689.1 | 14 | 2729 | AAH29689.1 |
More Information
Set Membership: |
|

Genomic Map