Assay Details
Target Gene Details
Entrez Gene ID: | 69034 |
Gene Name: | nuclear protein transcriptional regulator 1 like |
Gene Aliases: |
1810010E01Rik, 4930579G22Rik |
Location: |
Chr.5:129908540-129913789 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nupr1l | NM_026916.3 | 1 | 64 | NP_081192.2 |
| AK007417.1 | BAB25026.1 | |||
| AK016320.1 | 1 | 62 | ||
| AK135384.1 | BAE22513.1 | |||
| AK148362.1 | ||||
| CJ051706.1 |

Genomic Map