Assay Details
Target Gene Details
Entrez Gene ID: | 231252 |
Gene Name: | cholinergic receptor, nicotinic, alpha polypeptide 9 |
Gene Aliases: |
2410015I05Rik, Acra9, EG666827, Gm8311 |
Location: |
Chr.5:65934764-65977560 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrna9 | NM_001081104.1 | 5 | 1348 | NP_001074573.1 |
| AK010496.1 | 5 | 1341 | ||
| AY095483.1 | 2 | 518 | AAM34452.1 |
More Information
Set Membership: |
|

Genomic Map