Assay Details
Target Gene Details
Entrez Gene ID: | 231252 |
Gene Name: | cholinergic receptor, nicotinic, alpha polypeptide 9 |
Gene Aliases: |
2410015I05Rik, Acra9, EG666827, Gm8311 |
Location: |
Chr.5:65934764-65977560 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrna9 |

Genomic Map