Assay Details
Target Gene Details
Entrez Gene ID: | 12896 |
Gene Name: | carnitine palmitoyltransferase 2 |
Gene Aliases: |
AI323697, CPTII |
Location: |
Chr.4:107903982-107923589 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpt2 | NM_009949.2 | 4 | 684 | NP_034079.2 |
| AK144441.1 | 4 | 684 | BAE25892.1 | |
| AK150163.1 | 4 | 656 | BAE29353.1 | |
| AK169038.1 | 4 | 581 | BAE40828.1 | |
| BC138514.1 | 4 | 619 | ||
| BC145859.1 | 4 | 619 | ||
| U01163.1 | 4 | 629 | AAA18921.1 |
More Information
Set Membership: |
|

Genomic Map