Assay Details
Target Gene Details
Entrez Gene ID: | 71287 |
Gene Name: | carboxypeptidase, vitellogenic-like |
Gene Aliases: |
4933436L16Rik, HVLP |
Location: |
Chr.6:53873279-53978673 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpvl | NM_001289713.1 | NP_001276642.1 | ||
| NM_001289714.1 | NP_001276643.1 | |||
| NM_027749.2 | 2 | 127 | NP_082025.1 | |
| AK017087.1 | BAB30589.1 | |||
| BC137839.1 | 2 | 116 | ||
| BC144966.1 |
More Information
Set Membership: |
|

Genomic Map