Assay Details
Target Gene Details
Entrez Gene ID: | 12922 |
Gene Name: | corticotropin releasing hormone receptor 2 |
Gene Aliases: |
CRF-R2, CRFR2alpha, CRFR2beta, Crfr2 |
Location: |
Chr.6:55090048-55133016 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crhr2 | NM_001288618.1 | 13 | 1559 | NP_001275547.1 |
| NM_001288619.1 | 12 | 1466 | NP_001275548.1 | |
| NM_001288620.1 | 11 | 1356 | NP_001275549.1 | |
| NM_009953.4 | 13 | 1556 | NP_034083.2 | |
| AK142261.1 | 13 | 1506 | BAE24999.1 | |
| AY445512.1 | AAS07021.1 | |||
| AY753668.1 | AAU94301.1 | |||
| BC137592.1 | 13 | 1442 | ||
| U19939.1 | 13 | 1512 | AAC52243.1 | |
| U21729.1 | 13 | 1510 | AAC52174.1 |
More Information
Set Membership: |
|

Genomic Map