Assay Details
Target Gene Details
Entrez Gene ID: | 14763 |
Gene Name: | G protein-coupled receptor 37 |
Gene Aliases: |
AI848630, Pael-R |
Location: |
Chr.6:25668523-25689980 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr37 | NM_010338.2 | 2 | 2538 | NP_034468.2 |
| AK032525.1 | 2 | 2374 | BAC27911.1 | |
| BC031918.1 | 2 | 2406 | AAH31918.1 |
More Information
Set Membership: |
|

Genomic Map