Assay Details
Target Gene Details
Entrez Gene ID: | 80885 |
Gene Name: | hydroxycarboxylic acid receptor 2 |
Gene Aliases: |
Gpr109a, Gpr109b, HM74, Niacr1, PUMA-G, Pumag, mHM74b |
Location: |
Chr.5:123863570-123865516 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hcar2 | NM_030701.3 | 1 | 971 | NP_109626.1 |
| AJ300198.1 | 1 | 951 | CAC17790.1 | |
| AK150795.1 | 1 | 953 | BAE29858.1 | |
| AK169956.1 | 1 | 955 | ||
| BC120715.1 | 1 | 941 | ||
| BC125296.1 | 1 | 941 |

Genomic Map