Assay Details
Target Gene Details
Entrez Gene ID: | 12020 |
Gene Name: | NK3 homeobox 2 |
Gene Aliases: |
Bapx1, NKX3.2, Nkx-3.2 |
Location: |
Chr.5:41761483-41764220 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx3-2 | NM_007524.3 | 1 | 126 | NP_031550.2 |
| BC145872.1 | 1 | 121 | ||
| BC145874.1 | 1 | 121 | ||
| U87957.1 | 1 | 126 | AAB47764.1 |
More Information
Set Membership: |
|

Genomic Map