Assay Details
Target Gene Details
Entrez Gene ID: | 108099 |
Gene Name: | protein kinase, AMP-activated, gamma 2 non-catalytic subunit |
Gene Aliases: |
2410051C13Rik, AAKG, AAKG2, AI854673, H91620p, WPWS |
Location: |
Chr.5:24862735-25101906 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkag2 | NM_145401.2 | 1 | 486 | NP_663376.2 |
| AK154296.1 | 1 | 487 | BAE32497.1 | |
| BC015283.1 | 1 | 274 | AAH15283.1 |
More Information
Set Membership: |
|

Genomic Map