Assay Details
Target Gene Details
Entrez Gene ID: | 213393 |
Gene Name: | RIKEN cDNA 8430408G22 gene |
Gene Aliases: |
Depp, Fseg |
Location: |
Chr.6:116650684-116652847 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 8430408G22Rik | NM_001166580.1 | 2 | 891 | NP_001160052.1 |
| NM_145980.2 | 2 | 894 | NP_666092.1 | |
| AK078806.1 | 2 | 896 | BAC37402.1 | |
| AK087284.1 | 2 | 892 | BAC39837.1 | |
| BB683812.1 | ||||
| BC031533.1 | 2 | 879 | AAH31533.1 | |
| BC058515.1 | 2 | 862 | AAH58515.1 |
Target Gene Details
Entrez Gene ID: | 213391 |
Gene Name: | Ras association (RalGDS/AF-6) domain family member 4 |
Gene Aliases: |
3830411C14Rik, AD037, AI586137 |
Location: |
Chr.6:116632994-116673836 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rassf4 | NM_178045.4 | NP_835146.3 | ||
| AK036528.1 | BAC29463.1 | |||
| AK053578.1 | ||||
| AK089460.1 | ||||
| AK139284.1 | BAE23943.1 | |||
| AK151561.1 | ||||
| AK152639.1 | ||||
| AK160584.1 | BAE35891.1 | |||
| AK164357.1 | BAE37757.1 | |||
| AK170955.1 | BAE42138.1 | |||
| AK171198.1 | BAE42308.1 | |||
| BC023245.1 | AAH23245.1 | |||
| BC060709.1 | AAH60709.1 | |||
| BY170346.1 |
More Information
Set Membership: |
|

Genomic Map