Assay Details
Target Gene Details
Entrez Gene ID: | 330323 |
Gene Name: | family with sequence similarity 188, member B |
Gene Aliases: |
C330043M08Rik, Gm1406 |
Location: |
Chr.6:55203347-55320222 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam188b | NM_001142781.1 | NP_001136253.1 | ||
| NM_177883.4 | 11 | 2116 | NP_808551.2 | |
| AK082855.1 | 11 | 2117 | BAC38655.1 | |
| AK142385.1 | BAE25052.1 | |||
| AK161304.1 |

Genomic Map