Assay Details
Target Gene Details
Entrez Gene ID: | 330323 |
Gene Name: | family with sequence similarity 188, member B |
Gene Aliases: |
C330043M08Rik, Gm1406 |
Location: |
Chr.6:55203347-55320222 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam188b | NM_001142781.1 | 5 | 916 | NP_001136253.1 |
| NM_177883.4 | 5 | 916 | NP_808551.2 | |
| AK082855.1 | 5 | 917 | BAC38655.1 | |
| AK142385.1 | 5 | 834 | BAE25052.1 | |
| AK161304.1 | 5 | 794 | ||
| BC118940.1 | 5 | 910 |

Genomic Map