Assay Details
Target Gene Details
Entrez Gene ID: | 56449 |
Gene Name: | Y box protein 3 |
Gene Aliases: |
Csda, Dpba, MSY3, MSY4, Yb2, dbpA, oxyR |
Location: |
Chr.6:131364858-131388450 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ybx3 | NM_011733.2 | 9 | 1287 | NP_035863.1 |
| NM_139117.2 | 10 | 1494 | NP_620817.2 | |
| AF246224.1 | 10 | 1336 | AAF61741.1 | |
| AF248546.1 | 10 | 1316 | AAG14418.1 | |
| AF248547.1 | 9 | 1109 | AAG14419.1 | |
| AK003145.1 | 9 | 1218 | ||
| AK076041.1 | 10 | 1433 | BAC36141.1 | |
| BC027785.1 | 9 | 1297 | AAH27785.1 | |
| BC048242.1 | 10 | 1482 | AAH48242.1 | |
| BC062377.1 | 9 | 1275 | AAH62377.1 | |
| D14485.1 | 9 | 1273 | BAA03376.1 |

Genomic Map