Assay Details
Target Gene Details
Entrez Gene ID: | 66117 |
Gene Name: | RIKEN cDNA 1110001J03 gene |
Gene Aliases: |
Fmc1 |
Location: |
Chr.6:38534861-38539449 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110001J03Rik | NM_025363.3 | 2 | 336 | NP_079639.1 |
| AK003231.2 | 2 | 357 | BAC25025.1 | |
| AK008291.2 | 2 | 337 | BAB25581.1 | |
| AK009186.1 | 2 | 330 | BAB26127.1 | |
| BC059710.1 | 2 | 352 | AAH59710.1 |

Genomic Map