Assay Details
Target Gene Details
Entrez Gene ID: | 93746 |
Gene Name: | G protein-coupled receptor, family C, group 5, member D |
Gene Aliases: |
- |
Location: |
Chr.6:135105991-135137443 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gprc5d | NM_001205396.1 | 4 | 1299 | NP_001192325.1 |
| NM_053118.1 | 3 | 1231 | NP_444348.1 | |
| AB025221.1 | 3 | 1145 | BAC20266.1 | |
| AB025222.1 | 2 | 1077 | BAC20267.1 | |
| AB099816.1 | 2 | 1122 | BAC79168.1 | |
| AF218809.1 | 3 | 1231 | AAF72880.1 | |
| AK137739.1 | 4 | 1262 | BAE23485.1 |
More Information
Set Membership: |
|

Genomic Map