Assay Details
Target Gene Details
Entrez Gene ID: | 17057 |
Gene Name: | killer cell lectin-like receptor subfamily B member 1A |
Gene Aliases: |
Ly55a, NKR-P1.7, NKR-P1A, NKRP12, Nkrp1-a, Nkrp1a, ly-55A |
Location: |
Chr.6:128609227-128622985 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klrb1a | NM_001159902.1 | 5 | 862 | NP_001153374.1 |
| NM_010737.3 | 6 | 961 | NP_034867.3 | |
| BC120707.1 | 6 | 899 | ||
| BC145045.1 | 5 | 800 | ||
| M77753.1 | 6 | 860 | AAA39366.1 |
More Information
Set Membership: |
|

Genomic Map