Assay Details
Target Gene Details
Entrez Gene ID: | 80782 |
Gene Name: | killer cell lectin-like receptor subfamily B member 1B |
Gene Aliases: |
Klrb1d, Ly55b, Ly55d, NKR-P1B, NKR-P1D, Nkrp1-b, Nkrp1d |
Location: |
Chr.6:128813706-128826375 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klrb1b | NM_030599.4 | 6 | 1138 | NP_085102.4 |
| AF342896.1 | 6 | 1117 | AAK26639.1 | |
| AK030702.1 | 6 | 1087 | BAC27088.1 | |
| AK138766.1 | 5 | 1021 | ||
| AK169820.1 | 6 | 1155 | BAE41390.1 | |
| BC055379.1 | 6 | 1137 | AAH55379.1 | |
| BC100580.1 | 6 | 1125 | AAI00581.1 | |
| M77677.1 | 6 | 1114 | AAA39823.1 |
More Information
Set Membership: |
|

Genomic Map