Assay Details
Target Gene Details
Entrez Gene ID: | 74253 |
Gene Name: | killer cell lectin-like receptor subfamily G, member 2 |
Gene Aliases: |
2310020F24Rik |
Location: |
Chr.6:38621414-38637706 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klrg2 | NM_001033171.2 | 1 | 583 | NP_001028343.2 |
| AK009417.1 | 1 | 588 | ||
| AK145064.1 | 1 | 578 | BAE26215.1 | |
| AK162933.1 | ||||
| AK169862.1 | BAE41418.1 |
More Information
Set Membership: |
|

Genomic Map