Assay Details
Target Gene Details
Entrez Gene ID: | 330052 |
Gene Name: | RIKEN cDNA A930003O13 gene |
Gene Aliases: |
- |
Location: |
Chr.5:22738910-22746884 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A930003O13Rik | NR_027362.1 | 1 | 335 | |
| AK044256.1 | 1 | 359 |
Target Gene Details
Entrez Gene ID: | 269629 |
Gene Name: | lipoma HMGIC fusion partner-like 3 |
Gene Aliases: |
A930031L14Rik |
Location: |
Chr.5:22745636-23275597 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lhfpl3 | NM_001081231.2 | 1 | 358 | NP_001074700.2 |
| NM_029990.1 | 1 | 358 | NP_084266.1 | |
| AK020916.1 | 1 | 327 | BAB32253.1 | |
| AK033906.1 | 1 | 332 |

Genomic Map