Assay Details
Target Gene Details
Entrez Gene ID: | 269629 |
Gene Name: | lipoma HMGIC fusion partner-like 3 |
Gene Aliases: |
A930031L14Rik |
Location: |
Chr.5:22745636-23275597 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lhfpl3 | NM_001081231.2 | 4 | 1458 | NP_001074700.2 |
| NM_029990.1 | 3 | 1410 | NP_084266.1 | |
| AK033906.1 | 4 | 1432 | ||
| AK076730.1 | 3 | 1075 |

Genomic Map