Assay Details
Target Gene Details
Entrez Gene ID: | 67151 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, non-ATPase, 9 |
Gene Aliases: |
1500011J20Rik, 2610202L11Rik, Bridge-1, P27 |
Location: |
Chr.5:123228190-123250126 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmd9 | NM_026000.2 | 1 | 210 | NP_080276.2 |
| AK002280.1 | 1 | 134 | BAB21984.1 | |
| AK005216.1 | 1 | 143 | ||
| AK010276.1 | 1 | 98 | BAB26813.1 | |
| AK011863.1 | 1 | 143 | ||
| AK011984.1 | 1 | 155 | BAB27957.1 | |
| AK145487.1 | 1 | 155 | BAE26465.1 | |
| AK168339.1 | 1 | 133 | BAE40276.1 | |
| AK168863.1 | 1 | 152 | BAE40682.1 | |
| BC051930.1 | 1 | 95 | AAH51930.1 | |
| BC060245.1 | 1 | 138 | AAH60245.1 | |
| BY128235.1 | 1 | 211 |
More Information
Set Membership: |
|

Genomic Map